Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGACGCTCCGCAGTCCCGGGCAG[G/T]GGGCGGAGGGGGCGCCGGGGGAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609117 | ||||||||||||||||||||
Literature Links: |
FBXO46 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO46 - F-box protein 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080469.1 | 1186 | Missense Mutation | ACT,CCT | T,P 355 | NP_001073938.1 |
QPCTL - glutaminyl-peptide cyclotransferase like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |