Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTTAACAACACTTTGCACCTCA[T/C]TGGAGAGCACCATGATGGGGTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604955 MIM: 604946 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIR2DS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIR2DS4 - killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281971.1 | 307 | Missense Mutation | ACT,ATT | T,I 73 | NP_001268900.1 | |
NM_001281972.1 | 307 | Missense Mutation | ACT,ATT | T,I 73 | NP_001268901.1 | |
NM_012314.5 | 307 | Missense Mutation | ACT,ATT | T,I 73 | NP_036446.3 |
KIR3DL1 - killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |