Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGTGATGCCCACCACGGGCACC[C/T]GGGGCATCGGTGGGTAGCCCAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604018 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUMBL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUMBL - NUMB like, endocytic adaptor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289979.1 | 1601 | Missense Mutation | CAG,CGG | Q,R 463 | NP_001276908.1 | |
NM_001289980.1 | 1601 | Missense Mutation | CAG,CGG | Q,R 463 | NP_001276909.1 | |
NM_004756.4 | 1601 | Missense Mutation | CAG,CGG | Q,R 504 | NP_004747.1 |