Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACGCATCGGTCGCCTGGTCCTG[C/T]GCGCCTGCATGGAGAAGGGTGTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609169 MIM: 609969 MIM: 613585 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAPDHS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAPDHS - glyceraldehyde-3-phosphate dehydrogenase, spermatogenic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014364.4 | 286 | Missense Mutation | CGC,TGC | R,C 92 | NP_055179.1 | |
XM_017026579.1 | 286 | Missense Mutation | CGC,TGC | R,C 91 | XP_016882068.1 |
SBSN - suprabasin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM147 - transmembrane protein 147 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM147-AS1 - TMEM147 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |