Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGCCAGTGGGGCCGCGCTGGC[C/T]GCCACCATCCACTGCTATTGTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612685 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMSAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMSAP3 - calmodulin regulated spectrin associated protein family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080429.2 | 872 | Silent Mutation | GCC,GCT | A,A 266 | NP_001073898.1 | |
NM_020902.1 | 872 | Silent Mutation | GCC,GCT | A,A 239 | NP_065953.1 | |
XM_005272491.3 | 872 | Silent Mutation | GCC,GCT | A,A 255 | XP_005272548.1 | |
XM_011528154.2 | 872 | Silent Mutation | GCC,GCT | A,A 266 | XP_011526456.1 | |
XM_017027036.1 | 872 | Silent Mutation | GCC,GCT | A,A 266 | XP_016882525.1 |
MIR6792 - microRNA 6792 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |