Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGCTACTGGCGTTTTGTGTGGA[C/T]CATGCTCTCCCAGCCCCTTGTGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603823 | ||||||||||||||||||||
Literature Links: |
FFAR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FFAR2 - free fatty acid receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005306.2 | 614 | Missense Mutation | ACC,ATC | T,I 205 | NP_005297.1 | |
XM_017026709.1 | 614 | Missense Mutation | ACC,ATC | T,I 205 | XP_016882198.1 | |
XM_017026710.1 | 614 | Missense Mutation | ACC,ATC | T,I 205 | XP_016882199.1 | |
XM_017026711.1 | 614 | Missense Mutation | ACC,ATC | T,I 205 | XP_016882200.1 |