Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTGGGCACCTGAATGTAGCCCT[G/T]CTGGTCAATGAGCAGATTCTCCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601639 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKACA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKACA - protein kinase cAMP-activated catalytic subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304349.1 | 1166 | Missense Mutation | AAG,CAG | K,Q 254 | NP_001291278.1 | |
NM_002730.3 | 1166 | Missense Mutation | AAG,CAG | K,Q 178 | NP_002721.1 | |
NM_207518.2 | 1166 | Intron | NP_997401.1 | |||
XM_017026948.1 | 1166 | Intron | XP_016882437.1 |
SAMD1 - sterile alpha motif domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |