Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTGCCGCTTTGACTTCCCGGAT[A/G]AGCTGCGGCCCGCTGTGGTGCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 159460 | ||||||||||||||||||||
Literature Links: |
MAG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAG - myelin associated glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199216.1 | 343 | Missense Mutation | AAG,GAG | K,E 24 | NP_001186145.1 | |
NM_002361.3 | 343 | Missense Mutation | AAG,GAG | K,E 49 | NP_002352.1 | |
NM_080600.2 | 343 | Missense Mutation | AAG,GAG | K,E 49 | NP_542167.1 |