Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCAGACTGGGACCTCACCTCCC[C/T]AGCCAGCAAGGGGATTCCCCCAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605631 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAPSA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAPSA - napsin A aspartic peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004851.2 | 1466 | Missense Mutation | AGG,GGG | R,G 311 | NP_004842.1 | |
XM_011527540.1 | 1466 | Missense Mutation | AGG,GGG | R,G 311 | XP_011525842.1 | |
XM_017027512.1 | 1466 | Missense Mutation | AGG,GGG | R,G 302 | XP_016883001.1 |