Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAAAAGGTTGTACCAGTAGAAG[A/G]TTATGGAAGATTTTGTCATTGACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613894 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPY19L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPY19L3 - dpy-19 like 3 (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172774.1 | 299 | Silent Mutation | AGA,AGG | R,R 40 | NP_001166245.1 | |
NM_207325.2 | 299 | Silent Mutation | AGA,AGG | R,R 40 | NP_997208.2 | |
XM_006723045.1 | 299 | Intron | XP_006723108.1 | |||
XM_011526526.2 | 299 | Silent Mutation | AGA,AGG | R,R 40 | XP_011524828.1 | |
XM_017026366.1 | 299 | Silent Mutation | AGA,AGG | R,R 40 | XP_016881855.1 | |
XM_017026367.1 | 299 | UTR 5 | XP_016881856.1 |
LOC400684 - uncharacterized LOC400684 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |