Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCGCAGGTAGGACAGTAGGTCC[A/G]CTCGCAGCCTTGTCAGCACACCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147681 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM71E2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM71E2 - family with sequence similarity 71 member E2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL11 - interleukin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000641.3 | 449 | Missense Mutation | GCG,GTG | A,V 99 | NP_000632.1 | |
NM_001267718.1 | 449 | Missense Mutation | GCG,GTG | A,V 20 | NP_001254647.1 |
TMEM190 - transmembrane protein 190 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |