Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGAAGCGGCGGAACCGGCACAG[C/T]CTAGTGGGGCCTCAGCAGCTAGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606214 | ||||||||||||||||||||
Literature Links: |
SHKBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHKBP1 - SH3KBP1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138392.3 | 481 | Silent Mutation | AGC,AGT | S,S 144 | NP_612401.2 | |
XM_006723474.1 | 481 | Silent Mutation | AGC,AGT | S,S 144 | XP_006723537.1 | |
XM_006723475.1 | 481 | Silent Mutation | AGC,AGT | S,S 144 | XP_006723538.1 | |
XM_011527492.1 | 481 | Silent Mutation | AGC,AGT | S,S 144 | XP_011525794.1 | |
XM_011527493.1 | 481 | UTR 5 | XP_011525795.1 | |||
XM_017027473.1 | 481 | Silent Mutation | AGC,AGT | S,S 144 | XP_016882962.1 | |
XM_017027474.1 | 481 | UTR 5 | XP_016882963.1 |
SPTBN4 - spectrin beta, non-erythrocytic 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |