Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGAAGACGTGCACGATGTGCGG[G/T]TGTCGCACGCCCCGCAGGATGGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614997 MIM: 609435 MIM: 610712 | ||||||||||||||||||||
Literature Links: |
GATAD2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GATAD2A - GATA zinc finger domain containing 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA13 - NADH:ubiquinone oxidoreductase subunit A13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015965.6 | 440 | Intron | NP_057049.5 |
TSSK6 - testis specific serine kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032037.3 | 440 | Missense Mutation | CAA,CAC | Q,H 69 | NP_114426.1 |