Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGACCTGAGTGCCCAGCAGAT[A/G]GAGGAGCGCACCAGGGAGCTCATCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603419 MIM: 601117 | ||||||||||||||||||||
Literature Links: |
SGTA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SGTA - small glutamine rich tetratricopeptide repeat containing alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THOP1 - thimet oligopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003249.4 | 302 | Missense Mutation | ATA,ATG | I,M 33 | NP_003240.1 | |
XM_011528228.2 | 302 | Intron | XP_011526530.1 |