Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGGTACTTTGGAAGGGACGCGC[A/G]GGCCAGACGCGCCCAGACGGCCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616582 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LSR - lipolysis stimulated lipoprotein receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260489.1 | 342 | Missense Mutation | CAG,CGG | Q,R 40 | NP_001247418.1 | |
NM_001260490.1 | 342 | Missense Mutation | CAG,CGG | Q,R 40 | NP_001247419.1 | |
NM_015925.6 | 342 | Missense Mutation | CAG,CGG | Q,R 40 | NP_057009.3 | |
NM_205834.3 | 342 | Missense Mutation | CAG,CGG | Q,R 40 | NP_991403.1 | |
NM_205835.3 | 342 | Missense Mutation | CAG,CGG | Q,R 40 | NP_991404.1 | |
XM_005258980.1 | 342 | Missense Mutation | CAG,CGG | Q,R 40 | XP_005259037.1 | |
XM_005258982.1 | 342 | Missense Mutation | CAG,CGG | Q,R 40 | XP_005259039.1 | |
XM_011527026.1 | 342 | Missense Mutation | CAG,CGG | Q,R 40 | XP_011525328.1 |