Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACACACCTGGCTCAGGGCTCC[C/T]GTGAGACCACGCCACGCAGGTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610336 MIM: 605217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD4C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2CD4C - C2 calcium dependent domain containing 4C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHC2 - SHC adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012435.2 | 1619 | Missense Mutation | CAG,CGG | Q,R 580 | NP_036567.2 | |
XM_011527893.2 | 1619 | Intron | XP_011526195.1 | |||
XM_011527894.2 | 1619 | Intron | XP_011526196.1 | |||
XM_011527895.2 | 1619 | Missense Mutation | CAG,CGG | Q,R 535 | XP_011526197.1 | |
XM_011527896.2 | 1619 | Intron | XP_011526198.1 | |||
XM_017026567.1 | 1619 | Intron | XP_016882056.1 |