Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGCCGGGCCAGGAAGCTGTGAG[C/T]CAGCAGCGCCTCCTTGGTGTGCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602335 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMP3 - epithelial membrane protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM143 - transmembrane protein 143 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303538.1 | 1326 | Missense Mutation | ACT,GCT | T,A 337 | NP_001290467.1 | |
NM_001303539.1 | 1326 | Missense Mutation | ACT,GCT | T,A 307 | NP_001290468.1 | |
NM_001303540.1 | 1326 | Missense Mutation | ACT,GCT | T,A 272 | NP_001290469.1 | |
NM_018273.3 | 1326 | Missense Mutation | ACT,GCT | T,A 372 | NP_060743.2 |