Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCATTGATAGAAAGGCCGGCCTT[C/G]ACCTGTGGGTGTTAAAGGTATGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610349 MIM: 609623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAMSTR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAMSTR - MEF2 activating motif and SAP domain containing transcriptional regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASIP1 - Ras interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017805.2 | 2397 | Missense Mutation | CAA,GAA | Q,E 792 | NP_060275.2 | |
XM_011527053.2 | 2397 | Missense Mutation | CAA,GAA | Q,E 893 | XP_011525355.2 | |
XM_011527054.2 | 2397 | Missense Mutation | CAA,GAA | Q,E 893 | XP_011525356.2 | |
XM_011527055.2 | 2397 | Missense Mutation | CAA,GAA | Q,E 795 | XP_011525357.2 | |
XM_017026914.1 | 2397 | Missense Mutation | CAA,GAA | Q,E 893 | XP_016882403.1 | |
XM_017026915.1 | 2397 | Missense Mutation | CAA,GAA | Q,E 795 | XP_016882404.1 |