Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCGACAGCGGCTTGGGGTGCCA[C/G]GACACGTTGATGCTCATTGAGAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162860 MIM: 605157 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD177 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD177 - CD177 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020406.3 | 224 | Missense Mutation | CAC,CAG | H,Q 56 | NP_065139.2 | |
XM_017027021.1 | 224 | Missense Mutation | CAC,CAG | H,Q 56 | XP_016882510.1 | |
XM_017027022.1 | 224 | Missense Mutation | CAC,CAG | H,Q 56 | XP_016882511.1 |
PRG1 - p53-responsive gene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |