Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCAGGACAACCAAGGAAGTCCC[C/T]GCTCTGGATGCCGCGTGCTGTCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610477 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPRSS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMPRSS9 - transmembrane protease, serine 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527978.2 | 318 | Silent Mutation | CCC,CCT | P,P 19 | XP_011526280.1 | |
XM_011527980.1 | 318 | Intron | XP_011526282.1 | |||
XM_011527982.2 | 318 | Intron | XP_011526284.1 |