Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATACATCTAGTCTCATTAAACAC[A/G]GGAGGATTCACACTGGAGAGAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF586 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF586 - zinc finger protein 586 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077426.2 | 864 | Silent Mutation | ACA,ACG | T,T 182 | NP_001070894.1 | |
NM_001204814.1 | 864 | Missense Mutation | AGG,GGG | R,G 182 | NP_001191743.1 | |
NM_017652.3 | 864 | Missense Mutation | AGG,GGG | R,G 225 | NP_060122.2 |