Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCGCCAGCACCCAGGACCTGAG[C/T]CCCCACTAACGTCCAGTGAACTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126340 MIM: 601334 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERCC2 - ERCC excision repair 2, TFIIH core complex helicase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000400.3 | 2593 | Intron | NP_000391.1 | |||
NM_001130867.1 | 2593 | Intron | NP_001124339.1 | |||
XM_011526611.2 | 2593 | UTR 3 | XP_011524913.1 | |||
XM_017026467.1 | 2593 | UTR 3 | XP_016881956.1 |
KLC3 - kinesin light chain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177417.2 | 2593 | Silent Mutation | AGC,AGT | S,S 502 | NP_803136.2 |