Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCCCTCCGCGTGCTGCTGCTC[C/T]TCCAGGTCCCCCCCGGTGGCTCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614566 MIM: 191044 | ||||||||||||||||||||
Literature Links: |
DNAAF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAAF3 - dynein axonemal assembly factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256714.1 | 1220 | Silent Mutation | GAA,GAG | E,E 403 | NP_001243643.1 | |
NM_001256715.1 | 1220 | Silent Mutation | GAA,GAG | E,E 335 | NP_001243644.1 | |
NM_001256716.1 | 1220 | Silent Mutation | GAA,GAG | E,E 281 | NP_001243645.1 | |
NM_178837.4 | 1220 | Silent Mutation | GAA,GAG | E,E 382 | NP_849159.2 |
TNNI3 - troponin I3, cardiac type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |