Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCTTGGCTGCCTGCTCTGGGGT[A/G]GAGAACCGATTGTTCCGGGATTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600696 MIM: 603083 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ECH1 - enoyl-CoA hydratase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HNRNPL - heterogeneous nuclear ribonucleoprotein L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005335.1 | Intron | NP_001005335.1 | ||||
NM_001533.2 | Intron | NP_001524.2 | ||||
XM_011526886.1 | Intron | XP_011525188.1 | ||||
XM_011526887.1 | Intron | XP_011525189.1 | ||||
XM_011526888.2 | Intron | XP_011525190.1 | ||||
XM_011526889.1 | Intron | XP_011525191.1 |