Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGCCGGCCGTGCGCTGGGGCGG[G/T]TCCTCCACACTGATCACAGAGAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 130410 | ||||||||||||||||||||
Literature Links: |
ETFB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ETFB - electron transfer flavoprotein beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014763.1 | 779 | Missense Mutation | GAA,GAC | E,D 320 | NP_001014763.1 | |
NM_001985.2 | 779 | Missense Mutation | GAA,GAC | E,D 229 | NP_001976.1 |
VSIG10L - V-set and immunoglobulin domain containing 10 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |