Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGTCCCTGGATGGCTTTGTGTTC[A/G]CCTTGAACCAGGAAGGAAAATTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPAS1 - neuronal PAS domain protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321086.1 | 674 | Intron | NP_001308015.1 | |||
NM_002517.3 | 674 | Missense Mutation | ACC,GCC | T,A 152 | NP_002508.2 | |
XM_006723231.2 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_006723294.1 | |
XM_011526997.1 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_011525299.1 | |
XM_011526999.1 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_011525301.1 | |
XM_011527001.1 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_011525303.1 | |
XM_011527002.1 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_011525304.1 | |
XM_011527006.1 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_011525308.1 | |
XM_011527007.1 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_011525309.1 | |
XM_017026841.1 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_016882330.1 | |
XM_017026842.1 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_016882331.1 | |
XM_017026843.1 | 674 | Missense Mutation | ACC,GCC | T,A 152 | XP_016882332.1 | |
XM_017026844.1 | 674 | Missense Mutation | CAC,CGC | H,R 112 | XP_016882333.1 |