Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCATAGGCAGTTGGGCTTCCAG[A/G]CTGGTCTCCGGCAGAGGAGGATGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606475 MIM: 602139 | ||||||||||||||||||||
Literature Links: |
CD320 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD320 - CD320 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165895.1 | 733 | Missense Mutation | CCT,TCT | P,S 181 | NP_001159367.1 | |
NM_016579.3 | 733 | Missense Mutation | CCT,TCT | P,S 223 | NP_057663.1 |
NDUFA7 - NADH:ubiquinone oxidoreductase subunit A7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |