Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTCCTACTGCAGCAGGAGTTTTC[A/G]GTTTTGTCTGTATGTACAAGCCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IZUMO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IZUMO2 - IZUMO family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321449.1 | 863 | UTR 3 | NP_001308378.1 | |||
NM_152358.2 | 863 | Nonsense Mutation | CGA,TGA | R,* 216 | NP_689571.2 | |
XM_011526436.2 | 863 | Missense Mutation | CCG,CTG | P,L 206 | XP_011524738.1 | |
XM_011526437.2 | 863 | Missense Mutation | CCG,CTG | P,L 199 | XP_011524739.1 |