Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTGCCCGTGGGTTAGACTCGG[C/T]GAAGCAGGACAAGTAGAGGTTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176393 MIM: 176394 | ||||||||||||||||||||
Literature Links: |
PSG4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSG4 - pregnancy specific beta-1-glycoprotein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276495.1 | 1025 | Missense Mutation | ACC,GCC | T,A 263 | NP_001263424.1 | |
NM_001316339.1 | 1025 | Missense Mutation | ACC,GCC | T,A 234 | NP_001303268.1 | |
NM_002780.4 | 1025 | Missense Mutation | ACC,GCC | T,A 356 | NP_002771.2 | |
NM_213633.2 | 1025 | Missense Mutation | ACC,GCC | T,A 263 | NP_998798.1 | |
XM_017026995.1 | 1025 | Intron | XP_016882484.1 | |||
XM_017026996.1 | 1025 | Missense Mutation | ACC,GCC | T,A 372 | XP_016882485.1 | |
XM_017026997.1 | 1025 | Missense Mutation | ACC,GCC | T,A 356 | XP_016882486.1 | |
XM_017026998.1 | 1025 | Intron | XP_016882487.1 | |||
XM_017026999.1 | 1025 | Intron | XP_016882488.1 | |||
XM_017027000.1 | 1025 | Missense Mutation | ACC,GCC | T,A 279 | XP_016882489.1 | |
XM_017027001.1 | 1025 | Intron | XP_016882490.1 | |||
XM_017027002.1 | 1025 | Intron | XP_016882491.1 |
PSG5 - pregnancy specific beta-1-glycoprotein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |