Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTTGGCTGGGTTCCAGGGCGGA[A/C]GGACGCCGGTGGTCTCTGGCCTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612256 MIM: 609553 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAST1 - microtubule associated serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014975.2 | 383 | Silent Mutation | GAA,GAC | E,D 85 | NP_055790.1 | |
XM_011527805.2 | 383 | Missense Mutation | GAA,GAC | E,D 81 | XP_011526107.1 | |
XM_011527808.2 | 383 | Intron | XP_011526110.1 | |||
XM_017026501.1 | 383 | Intron | XP_016881990.1 |
MIR6794 - microRNA 6794 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTBDN - retbindin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |