Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGGTACTTCAATAGCCCCTAC[C/G]CCAAGAACTACCCCCCGGTGGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159460 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAG - myelin associated glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199216.1 | 394 | Missense Mutation | CCC,GCC | P,A 41 | NP_001186145.1 | |
NM_002361.3 | 394 | Missense Mutation | CCC,GCC | P,A 66 | NP_002352.1 | |
NM_080600.2 | 394 | Missense Mutation | CCC,GCC | P,A 66 | NP_542167.1 |