Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTTTGCGCACGGTGGTCTCGTC[C/G]AGGGAGAAGAGGTCGAAGTCGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159556 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLLT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MLLT1 - MLLT1, super elongation complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005934.3 | 1764 | Silent Mutation | CTC,CTG | L,L 542 | NP_005925.2 | |
XM_011528021.1 | 1764 | Silent Mutation | CTC,CTG | L,L 583 | XP_011526323.1 | |
XM_011528022.1 | 1764 | Silent Mutation | CTC,CTG | L,L 544 | XP_011526324.1 | |
XM_011528023.1 | 1764 | Silent Mutation | CTC,CTG | L,L 541 | XP_011526325.1 | |
XM_017026819.1 | 1764 | Silent Mutation | CTC,CTG | L,L 510 | XP_016882308.1 |