Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCGACCTGCACGCCCGATACGCT[C/G]AGTGTGGTTTGCGGATCCTGGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138322 MIM: 615729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPX4 - glutathione peroxidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039847.2 | 470 | Missense Mutation | CAG,GAG | Q,E 92 | NP_001034936.1 | |
NM_001039848.2 | 470 | Missense Mutation | CAG,GAG | Q,E 129 | NP_001034937.1 | |
NM_002085.4 | 470 | Missense Mutation | CAG,GAG | Q,E 92 | NP_002076.2 |
SBNO2 - strawberry notch homolog 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |