Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGACTTTAGACCGAAACTTGTCAA[C/T]GGGAAGGGTCCCTTAGATAACTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601246 MIM: 601768 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHAF1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHAF1A - chromatin assembly factor 1 subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005483.2 | 379 | Silent Mutation | AAC,AAT | N,N 93 | NP_005474.2 | |
XM_011527605.2 | 379 | Silent Mutation | AAC,AAT | N,N 93 | XP_011525907.1 | |
XM_011527606.2 | 379 | Silent Mutation | AAC,AAT | N,N 93 | XP_011525908.1 | |
XM_011527607.2 | 379 | Silent Mutation | AAC,AAT | N,N 93 | XP_011525909.1 |
SH3GL1 - SH3 domain containing GRB2 like 1, endophilin A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |