Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATATTCCTGATGCTCACATGATT[C/T]GTGTCCAGTGTCAACTCTGATGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF823 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF823 - zinc finger protein 823 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080493.3 | 433 | Missense Mutation | AAA,GAA | K,E 124 | NP_001073962.1 | |
NM_001297610.1 | 433 | UTR 5 | NP_001284539.1 | |||
NM_017507.1 | 433 | Missense Mutation | AAA,GAA | K,E 80 | NP_059977.1 | |
XM_017026941.1 | 433 | UTR 5 | XP_016882430.1 | |||
XM_017026942.1 | 433 | UTR 5 | XP_016882431.1 |