Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCCCCACTGCGGGTGCAGAGCA[C/T]GGTAAGTGTCAGGTGGCCCTTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605343 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSTL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FSTL3 - follistatin like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRSS57 - protease, serine 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308209.1 | 626 | Missense Mutation | ATG,GTG | M,V 199 | NP_001295138.1 | |
NM_214710.4 | 626 | Missense Mutation | ATG,GTG | M,V 200 | NP_999875.1 |