Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCCGGGAGGCTCCTCTGCACCA[G/T]CACAGAGCGCGAGCGGAAGTGGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601052 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OLFM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OLFM2 - olfactomedin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304347.1 | 1055 | Missense Mutation | ATG,CTG | M,L 323 | NP_001291276.1 | |
NM_001304348.1 | 1055 | Missense Mutation | ATG,CTG | M,L 221 | NP_001291277.1 | |
NM_058164.3 | 1055 | Missense Mutation | ATG,CTG | M,L 299 | NP_477512.1 |
PIN1 - peptidylprolyl cis/trans isomerase, NIMA-interacting 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |