Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGAATCACCATCAGTGGTGCCC[A/G]GTGACATGCTGGAGGAAGCTGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611712 MIM: 615698 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIPK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIPK4 - homeodomain interacting protein kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144685.4 | 2142 | Silent Mutation | ACC,ACT | T,T 613 | NP_653286.2 | |
XM_006723036.2 | 2142 | Silent Mutation | ACC,ACT | T,T 616 | XP_006723099.1 |
MIR6796 - microRNA 6796 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLD3 - phospholipase D family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |