Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGACCGGCTGCAGGGCCGAAG[C/G]GGGTGGTGACTCACGCTCCTTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615331 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRF2BP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRF2BP1 - interferon regulatory factor 2 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYPOP - Myb related transcription factor, partner of profilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012643.3 | 700 | Missense Mutation | CCT,GCT | P,A 205 | NP_001012661.1 | |
XM_017026742.1 | 700 | Missense Mutation | CCT,GCT | P,A 348 | XP_016882231.1 | |
XM_017026743.1 | 700 | Silent Mutation | CCC,CCG | P,P 384 | XP_016882232.1 |