Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCACATCTTGGTTTTCTGTGTG[A/G]GTCTCCTCACCATGGCCAAGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602359 MIM: 606684 MIM: 608303 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FXYD1 - FXYD domain containing ion transport regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278717.1 | 126 | Missense Mutation | AGT,GGT | S,G 13 | NP_001265646.1 | |
NM_001278718.1 | 126 | Missense Mutation | AGT,GGT | S,G 13 | NP_001265647.1 | |
NM_005031.4 | 126 | Missense Mutation | AGT,GGT | S,G 13 | NP_005022.2 | |
NM_021902.3 | 126 | Missense Mutation | AGT,GGT | S,G 13 | NP_068702.1 | |
XM_017026874.1 | 126 | Missense Mutation | AGT,GGT | S,G 13 | XP_016882363.1 | |
XM_017026875.1 | 126 | Missense Mutation | AGT,GGT | S,G 31 | XP_016882364.1 | |
XM_017026876.1 | 126 | Missense Mutation | AGT,GGT | S,G 31 | XP_016882365.1 |
FXYD7 - FXYD domain containing ion transport regulator 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LGI4 - leucine rich repeat LGI family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |