Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCGCTGTCCTGCCCTCCCACAG[A/G]CCGCATCGACTGTCAGCACCGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607246 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP3D1 - adaptor related protein complex 3 delta 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IZUMO4 - IZUMO family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031735.2 | 469 | Missense Mutation | AAC,AGC | N,S 124 | NP_001026905.2 | |
NM_001039846.1 | 469 | Missense Mutation | AAC,AGC | N,S 124 | NP_001034935.1 | |
XM_005259480.3 | 469 | Missense Mutation | AAC,AGC | N,S 124 | XP_005259537.2 |
MOB3A - MOB kinase activator 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |