Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACGTGCTGGCCGAGTACCTCACC[A/G]TGGCCATGCTCATGATTGGGGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612891 MIM: 603014 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC8E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC8E - leucine rich repeat containing 8 family member E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001268284.2 | 341 | Missense Mutation | ATG,GTG | M,V 33 | NP_001255213.1 | |
NM_001268285.2 | 341 | Intron | NP_001255214.1 | |||
NM_025061.5 | 341 | Missense Mutation | ATG,GTG | M,V 33 | NP_079337.2 | |
XM_011528319.2 | 341 | Missense Mutation | ATG,GTG | M,V 33 | XP_011526621.1 |
MAP2K7 - mitogen-activated protein kinase kinase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |