Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGGGGAGTGCAGGAGGGCCGG[C/G]CCCATGGCTCCGGGCTGCTGGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615331 | ||||||||||||||||||||
Literature Links: |
IRF2BP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IRF2BP1 - interferon regulatory factor 2 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYPOP - Myb related transcription factor, partner of profilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012643.3 | 635 | Missense Mutation | GCC,GGC | A,G 183 | NP_001012661.1 | |
XM_017026742.1 | 635 | Missense Mutation | GCC,GGC | A,G 326 | XP_016882231.1 | |
XM_017026743.1 | 635 | Missense Mutation | CCC,GCC | P,A 363 | XP_016882232.1 |