Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCTTTTGTGTTTTCTTGTAGAA[A/G]GACAAGGCACTCTCTCCACAGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C1orf162 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf162 - chromosome 1 open reading frame 162 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300834.1 | 1562 | Missense Mutation | AGA,GGA | R,G 14 | NP_001287763.1 | |
NM_001300835.1 | 1562 | Silent Mutation | AAA,AAG | K,K 16 | NP_001287764.1 | |
NM_174896.3 | 1562 | Missense Mutation | AGA,GGA | R,G 14 | NP_777556.1 | |
XM_011540712.2 | 1562 | Missense Mutation | AGA,GGA | R,G 14 | XP_011539014.1 | |
XM_017000323.1 | 1562 | Missense Mutation | AGA,GGA | R,G 14 | XP_016855812.1 | |
XM_017000324.1 | 1562 | Silent Mutation | AAA,AAG | K,K 16 | XP_016855813.1 |
TMIGD3 - transmembrane and immunoglobulin domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |