Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGCGGTGGAGCCACAGTCCCTG[C/T]GGTCCCGGTATCCAGTCTGGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609183 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AURKAIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AURKAIP1 - aurora kinase A interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127229.1 | 448 | Missense Mutation | ACA,GCA | T,A 92 | NP_001120701.1 | |
NM_001127230.1 | 448 | Missense Mutation | ACA,GCA | T,A 92 | NP_001120702.1 | |
NM_017900.2 | 448 | Missense Mutation | ACA,GCA | T,A 92 | NP_060370.1 | |
XM_011541670.2 | 448 | Missense Mutation | ACA,GCA | T,A 121 | XP_011539972.1 |