Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCATCGGCACGTGTGCAACTCCA[C/G]GAGCCGCTGGCACTGCCGGCACTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603490 | ||||||||||||||||||||
Literature Links: |
WNT4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WNT4 - Wnt family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030761.4 | 1206 | Missense Mutation | CTG,GTG | L,V 345 | NP_110388.2 | |
XM_011541597.2 | 1206 | Missense Mutation | CTG,GTG | L,V 367 | XP_011539899.1 | |
XM_011541598.2 | 1206 | Missense Mutation | CTG,GTG | L,V 290 | XP_011539900.1 | |
XM_011541599.1 | 1206 | Intron | XP_011539901.1 |