Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTCCCTACTCCTAGGCTCACCA[C/T]TGATGAGGCCCCTCAGGAGAGGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604468 MIM: 608042 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K6 - mitogen-activated protein kinase kinase kinase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYTL1 - synaptotagmin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193308.1 | 619 | Missense Mutation | ACT,ATT | T,I 141 | NP_001180237.1 | |
NM_032872.2 | 619 | Missense Mutation | ACT,ATT | T,I 141 | NP_116261.1 | |
XM_005246022.3 | 619 | Missense Mutation | ACT,ATT | T,I 173 | XP_005246079.1 | |
XM_006710990.1 | 619 | Missense Mutation | ACT,ATT | T,I 173 | XP_006711053.1 | |
XM_006710991.1 | 619 | UTR 5 | XP_006711054.1 |