Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGGCGAGCGAGGGCGCCGAGCAG[C/T]CCCCGGACCTGCACTGCGTGCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126335 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GADD45A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GADD45A - growth arrest and DNA damage inducible alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199741.1 | 673 | Missense Mutation | CCC,TCC | P,S 85 | NP_001186670.1 | |
NM_001199742.1 | 673 | Intron | NP_001186671.1 | |||
NM_001924.3 | 673 | Missense Mutation | CCC,TCC | P,S 119 | NP_001915.1 |