Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTGGACATCTGCTTTACCACCA[C/G]TGTTGCCCCACAGTTGCTGGTTACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR2G6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR2G6 - olfactory receptor family 2 subfamily G member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013355.1 | 227 | Missense Mutation | ACT,AGT | T,S 76 | NP_001013373.1 |
OR2T10 - olfactory receptor family 2 subfamily T member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004693.1 | 227 | Intron | NP_001004693.1 | |||
XM_017000273.1 | 227 | Intron | XP_016855762.1 |