Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTGAGGCCCGTCTGGCCAAAGG[C/T]GCCCAGCTCAACGGCCGCGACGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606066 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHX9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LHX9 - LIM homeobox 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014434.1 | 893 | Silent Mutation | GGC,GGT | G,G 41 | NP_001014434.1 | |
NM_020204.2 | 893 | Silent Mutation | GGC,GGT | G,G 50 | NP_064589.2 | |
XM_005245350.3 | 893 | Silent Mutation | GGC,GGT | G,G 56 | XP_005245407.2 | |
XM_011509781.2 | 893 | Silent Mutation | GGC,GGT | G,G 56 | XP_011508083.2 | |
XM_017001849.1 | 893 | Silent Mutation | GGC,GGT | G,G 31 | XP_016857338.1 |